Anti-VPS28 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302924EA 456 EUR
ORIGTA302924
Anti-VPS28 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1-3µg/ml; Immunochemistry: 10µg/ml

Type: Primary
Antigen: VPS28 (vacuolar protein sorting 28 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR