Anti-MPG Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302936EA 500 EUR
ORIGTA302936
Anti-MPG Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MPG (N-methylpurine-DNA glycosylase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR