Anti-ABCC4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302970EA 547 EUR
ORIGTA302970
Anti-ABCC4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000 Western Blot: 1-3µg/ml; Immunochemistry: 2-4µg/ml

Type: Primary
Antigen: ABCC4 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR