Anti-HPGD Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302977EA 500 EUR
ORIGTA302977
Anti-HPGD Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000 Western Blot: 01-1µg/ml

Type: Primary
Antigen: HPGD (hydroxyprostaglandin dehydrogenase 15-(NAD))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR