Anti-SRD5A2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302992EA 550 EUR
ORIGTA302992
Anti-SRD5A2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-10 µg/ml

Type: Primary
Antigen: SRD5A2 (steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR