Anti-RARA Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303003EA 547 EUR
ORIGTA303003
Anti-RARA Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 003-03µg/ml

Type: Primary
Antigen: RARA (retinoic acid receptor, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR