Anti-CCKAR Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303030EA 547 EUR
ORIGTA303030
Anti-CCKAR Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000 Western Blot: 10µg/ml

Type: Primary
Antigen: CCKAR (cholecystokinin A receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR