Anti-EIF3M Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303063EA 547 EUR
ORIGTA303063
Anti-EIF3M Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: EIF3M (eukaryotic translation initiation factor 3, subunit M)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR