Anti-CBX1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303064EA 547 EUR
ORIGTA303064
Anti-CBX1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-10 µg/ml

Type: Primary
Antigen: CBX1 (chromobox homolog 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR