Anti-KPNB1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303073EA 547 EUR
ORIGTA303073
Anti-KPNB1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: KPNB1 (karyopherin (importin) beta 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR