Anti-VPS25 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303080EA 454 EUR
ORIGTA303080
Anti-VPS25 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:1000; Western Blot: 05-15 µg/ml; Immunochemistry: 10µg/ml

Type: Primary
Antigen: VPS25 (vacuolar protein sorting 25 homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR