Anti-ACSL5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303108EA 547 EUR
ORIGTA303108
Anti-ACSL5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ACSL5 (acyl-CoA synthetase long-chain family member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR