Anti-CHGA Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303114EA 456 EUR
ORIGTA303114
Anti-CHGA Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: CHGA (chromogranin A (parathyroid secretory protein 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR