Anti-TCFL5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303152EA 547 EUR
ORIGTA303152
Anti-TCFL5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: TCFL5 (transcription factor-like 5 (basic helix-loop-helix))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR