Anti-GFAP Goat polyclonal antibody

Supplier: OriGene
ORIGTA303157EA 456 EUR
ORIGTA303157
Anti-GFAP Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1?g/ml; Immunochemistry: 2-4?g/ml Assay: 5-10?g/ml

Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR