Anti-S100A4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303160EA 547 EUR
ORIGTA303160
Anti-S100A4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: S100A4 (S100 calcium binding protein A4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR