Anti-ICAM5 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303165EA 547 EUR
ORIGTA303165
Anti-ICAM5 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000 Western Blot: 2-4µg/ml

Type: Primary
Antigen: ICAM5 (intercellular adhesion molecule 5, telencephalin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR