Anti-PIK3CA Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303167EA 500 EUR
ORIGTA303167
Anti-PIK3CA Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000 Western Blot: 2-3µg/ml

Type: Primary
Antigen: PIK3CA (phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR