Anti-GJB2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303171EA 547 EUR
ORIGTA303171
Anti-GJB2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: GJB2 (gap junction protein, beta 2, 26kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR