Anti-DGAT1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303180EA 547 EUR
ORIGTA303180
Anti-DGAT1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-1µg/ml

Type: Primary
Antigen: DGAT1 (diacylglycerol O-acyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR