Anti-RNF213 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303190EA 547 EUR
ORIGTA303190
Anti-RNF213 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 25µg/ml

Type: Primary
Antigen: RNF213 (ring finger protein 213)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR