Anti-TBC1D10C Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303203EA 702 EUR
ORIGTA303203
Anti-TBC1D10C Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: TBC1D10C (TBC1 domain family, member 10C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR