Anti-PDIA2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303215EA 500 EUR
ORIGTA303215
Anti-PDIA2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: PDIA2 (protein disulfide isomerase family A, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR