Anti-FOXO1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303221EA 547 EUR
ORIGTA303221
Anti-FOXO1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: FOXO1 (forkhead box O1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR