Anti-APBA1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303223EA 500 EUR
ORIGTA303223
Anti-APBA1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: APBA1 (amyloid beta (A4) precursor protein-binding, family A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR