Anti-G6PD Goat polyclonal antibody

Supplier: OriGene
ORIGTA303241EA 550 EUR
ORIGTA303241
Anti-G6PD Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: G6PD (glucose-6-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR