Anti-IGFBP3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303275EA 547 EUR
ORIGTA303275
Anti-IGFBP3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 002-006µg/ml

Type: Primary
Antigen: IGFBP3 (insulin-like growth factor binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR