Anti-KCNJ6 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303283EA 500 EUR
ORIGTA303283
Anti-KCNJ6 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: KCNJ6 (potassium inwardly-rectifying channel, subfamily J, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR