Anti-SLC12A4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303291EA 500 EUR
ORIGTA303291
Anti-SLC12A4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: SLC12A4 (solute carrier family 12 (potassium/chloride transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR