Anti-ACVR1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303310EA 500 EUR
ORIGTA303310
Anti-ACVR1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: ACVR1 (activin A receptor, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR