Anti-NPRL2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303320EA 454 EUR
ORIGTA303320
Anti-NPRL2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0 1-03µg/ml

Type: Primary
Antigen: NPRL2 (nitrogen permease regulator-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR