Anti-MC2R Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303323EA 502 EUR
ORIGTA303323
Anti-MC2R Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: MC2R (melanocortin 2 receptor (adrenocorticotropic hormone))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR