Anti-MARCH6 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303324EA 500 EUR
ORIGTA303324
Anti-MARCH6 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 02-05µg/ml

Type: Primary
Antigen: MARCH6 (membrane-associated ring finger (C3HC4) 6, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR