Anti-APOBEC3C Goat polyclonal antibody

Supplier: OriGene
ORIGTA303327EA 500 EUR
ORIGTA303327
Anti-APOBEC3C Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:1000; Western Blot: 05-15µg/ml

Type: Primary
Antigen: APOBEC3C (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR