Anti-TOM1L1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303340EA 454 EUR
ORIGTA303340
Anti-TOM1L1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: TOM1L1 (target of myb1 (chicken)-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR