Anti-DACH2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA303341EA 454 EUR
ORIGTA303341
Anti-DACH2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: DACH2 (dachshund family transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR