Anti-IRAK3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303364EA 547 EUR
ORIGTA303364
Anti-IRAK3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.05-0.2?g/ml

Type: Primary
Antigen: IRAK3 (interleukin-1 receptor-associated kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR