Anti-CRKL Rabbit monoclonal antibody [clone: EP270Y]

Supplier: OriGene
ORIGTA303464EA 673 EUR
ORIGTA303464
Anti-CRKL Rabbit monoclonal antibody [clone: EP270Y]
Antibodies
Recommended Dilutions: Western Blot: 1:50000; Immunoprecipitation: 1:50

Type: Primary
Antigen: CRKL (v-crk avian sarcoma virus CT10 oncogene homolog-like)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR