Anti-ERBB3 Rabbit Monoclonal Antibody [clone: EPR2325]

Supplier: OriGene
ORIGTA303500EA 685 EUR
ORIGTA303500
Anti-ERBB3 Rabbit Monoclonal Antibody [clone: EPR2325]
Antibodies
Recommended Dilutions: Western Blot: 1:2500-1:5000; Immunochemistry/Immunofluorescence: 1:100

Type: Primary
Antigen: ERBB3 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR