Anti-IRS1 Rabbit Monoclonal Antibody [clone: EP260Y]

Supplier: OriGene
ORIGTA303552EA 643 EUR
ORIGTA303552
Anti-IRS1 Rabbit Monoclonal Antibody [clone: EP260Y]
Antibodies
Recommended Dilutions: Western Blot: 1:500000

Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR