Anti-MCAM Rabbit Monoclonal Antibody [clone: EPR3208]

Supplier: OriGene
ORIGTA303592EA 715 EUR
ORIGTA303592
Anti-MCAM Rabbit Monoclonal Antibody [clone: EPR3208]
Recommended Dilutions: Immunochemistry: 1:250; Western Blot: 1:10000-1:50000; Immunoprecipitation: 1:10;Immunochemistry: 1:100-1:500; Flow Cytometry: 1:80

Type: Primary
Antigen: MCAM (melanoma cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR