Anti-DYNLL1 Rabbit Monoclonal Antibody [clone: EP1660Y]

Supplier: OriGene
ORIGTA303752EA 788 EUR
ORIGTA303752
Anti-DYNLL1 Rabbit Monoclonal Antibody [clone: EP1660Y]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:100-1:250; Immunoprecipitation: 1:100; Immunochemistry/Immunofluorescence: 1:100-1:250

Type: Primary
Antigen: DYNLL1 (dynein, light chain, LC8-type 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR