Anti-MAPKAPK2 Rabbit Monoclonal Antibody [clone: E341]

Supplier: OriGene
ORIGTA303762EA 664 EUR
ORIGTA303762
Anti-MAPKAPK2 Rabbit Monoclonal Antibody [clone: E341]
Antibodies
Recommended Dilutions: Western Blot: 1:500-1000; Immunoprecipitation: 1:200

Type: Primary
Antigen: MAPKAPK2 (mitogen-activated protein kinase-activated protein kinase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR