Anti-NEFM Rabbit Monoclonal Antibody [clone: EPR580(2)Y]

Supplier: OriGene
ORIGTA303832EA 566 EUR
ORIGTA303832
Anti-NEFM Rabbit Monoclonal Antibody [clone: EPR580(2)Y]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:100-250; Immunoprecipitation: 1:30

Type: Primary
Antigen: NEFM (neurofilament, medium polypeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR