Anti-TSC2 Rabbit Monoclonal Antibody [clone: EP1062Y]

Supplier: OriGene
ORIGTA303844EA 633 EUR
ORIGTA303844
Anti-TSC2 Rabbit Monoclonal Antibody [clone: EP1062Y]
Antibodies
Recommended Dilutions: Western Blot: 1:10000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR