Anti-GCH1 Goat antibody

Supplier: OriGene
ORIGTA305613EA 454 EUR
ORIGTA305613
Anti-GCH1 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 05-15?g/ml

Type: Primary
Antigen: GCH1 (GTP cyclohydrolase 1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR