Anti-CYBB Goat Antibody

Supplier: OriGene
ORIGTA305618EA 454 EUR
ORIGTA305618
Anti-CYBB Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: CYBB (cytochrome b-245, beta polypeptide)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR