Anti-ALOX15 Goat Antibody

Supplier: OriGene
ORIGTA305627EA 454 EUR
ORIGTA305627
Anti-ALOX15 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 02-05?g/ml

Type: Primary
Antigen: ALOX15 (arachidonate 15-lipoxygenase)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR