Anti-DKC1 Goat antibody

Supplier: OriGene
ORIGTA305629EA 547 EUR
ORIGTA305629
Anti-DKC1 Goat antibody
Antibodies
Recommended Dilutions: Western Blot: 001?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: DKC1 (dyskeratosis congenita 1, dyskerin)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR