Anti-ALOX5AP Goat Antibody

Supplier: OriGene
ORIGTA305631EA 550 EUR
ORIGTA305631
Anti-ALOX5AP Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR